missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalates Mixture 576 1000-5000 μg/mL in Dichloromethane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Dibutyl phthalate∣Diisononyl Phthalate (mixture of branched chain isomers)∣Di-n-octyl phthalate∣Phthalic acid, benzylbutyl ester∣Phthalic acid, bis-2-ethylhexyl ester∣Phthalic acid, bis-hexyl ester∣Phthalic acid, bis-iso-butyl ester∣Phthalic acid, bis-isodecyl ester |
| CAS | 84-74-2, 68515-48-0, 117-84-0, 85-68-7, 117-81-7, 84-75-3 |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Dibutyl phthalate 1000 μg/mL∣Di-n-octyl phthalate 1000 μg/mL∣Phthalic acid, benzylbutyl ester 1000 μg/mL∣Phthalic acid, bis-2-ethylhexyl ester 1000 μg/mL∣Phthalic acid, bis-hexyl ester 1000 μg/mL∣Phthalic acid, bis-iso-butyl ester 1000 μg/mL∣Diisononyl Phthalate (mixture of branched chain isomers) 5000 μg/mL∣Phthalic acid, bis-isodecyl ester 5000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?