Learn More
1000ppm of Phthalate Standard in Isooctane, 1 mL Ampule, Six Component Phthalate Standard, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Brand: SPEX Certiprep C1001-09
Code : NEW
Additional Details : CAS Number : 26761-40-0 Weight : 0.00100kg Transport : UN number : 1262 Chem class : 3 Pack group : II
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Bis(2-Ethylhexyl)phthalate | 117-81-7 | 1000μg/mL |
Butylbenzyl phthalate | 85-68-7 | 1000μg/mL |
Diisodecyl phthalate | 26761-40-0 | 1000μg/mL |
Diisononyl phthalate | 28553-12-0 | 1000μg/mL |
Di-n-butyl phthalate | 84-74-2 | 1000μg/mL |
Di-n-octyl phthalate | 117-84-0 | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Phthalate Standard | |
0.1 | |
1000μg/mL | |
bis(2-Ethylhexyl)phthalate,Butylbenzyl phthalate,Diisodecyl phthalate | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Six components | |
68515-48-0,26761-40-0,117-84-0,540-84-1,117-81-7, 7440-48-4 | |
2mL amber prescored glass ampule | |
Isooctane | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
Safety and Handling
- Danger
Hazard Category
- Aspiration toxicity Category 1
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P321-Specific treatment (see on this label).
- P331-Do NOT induce vomiting.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2,2,4-trimethylpentane, dibutyl phthalate, BBP, bis(2-ethylhexyl) phthalate, di-n-octyl phthalate
Product intended for laboratory use only