Product Code. 16399827
Shop All SPEX Certiprep Products
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
16399827
1 mL
1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code.
16399827
Supplier
SPEX Certiprep
Supplier No.
8061X
£149.00 £117.99 / 1mL Save £31.01 20% Off
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Includes
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte CAS No. Concentration Bis(2-Ethylhexyl)phthalate 117-81-7 2000μg/mL Bis(2-methoxyethyl)phthalate 117-82-8 2000μg/mL Bis(2-n-butoxyethyl)phthalate 117-83-9 2000μg/mL Bis(4-methyl-2-pentyl)phthalate 146-50-9 2000μg/mL Butylbenzyl phthalate 85-68-7 2000μg/mL Diamyl phthalate 131-18-0
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Chemical Name or Material
Phthalate Esters Mix
Packaging
2mL amber prescored glass ampule
Concentration or Composition Notes
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
CAS Max %
97.0, 0.2, 0.2, 0.2, 0.2, 0.2
CAS
84-61-7, 117-83-9, 84-76-4, 146-50-9, 84-75-3
Methods
For US EPA Methods 625, 8270C
Analyte or Component Names
bis(2-Ethylhexyl)phthalate,Bis(2-methoxyethyl )phthalate,Bis(2-n-butoxyethyl)phthalate
Technique
AA, ICP
For Use With (Equipment)
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Type
Certified Reference Material
Element
bis(2-Ethylhexyl)phthalate, bis(2-methoxyethyl )phthalate, bis(2-n-butoxyethyl)phthalate
Solvent or Matrix
Hexane
Quantity
1 mL
Concentration or Composition (by Analyte or Components)
2000μg/mL
Show More
Show Less
Product Identifier
Signal Word
Hazard Category
Aspiration toxicity Category 1
Carcinogenicity Category 2
Flammable liquid Category 2
Reproductive toxicity Category 1A
Skin corrosion/irritation Category 2
Skin sensitiser Category 1
Specific target organ toxicity after repeated exposure Category 2
Specific target organ toxicity Category 3
Hazard Statement
H225-Highly flammable liquid and vapour.
H304-May be fatal if swallowed and enters airways.
H315-Causes skin irritation.
H317-May cause an allergic skin reaction.
H336-May cause drowsiness or dizziness.
H351-Suspected of causing cancer.
H360-May damage fertility or the unborn child.
H373-May cause damage to organs.
Precautionary Statement
P101-If medical advice is needed, have product container or label at hand.
P102-Keep out of reach of children.
P103-Read label before use.
P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
P321-Specific treatment (see on this label).
P405-Store locked up.
P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
MIXTURE LIST-Contain: n-hexane, diisobutylphthalate, BBP
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?