missing translation for 'onlineSavingsMsg'
Learn More

Phthalanilic Acid (~90%), TRC

Product Code. 30135829
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135829 10 g 10g
30134803 5 g 5g
17961255 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135829 Supplier Toronto Research Chemicals Supplier No. TRCP40075810G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4727-29-1
Molecular Formula C14 H11 N O3
Molecular Weight (g/mol) 241.24
Synonym Benzoic acid, 2-[(phenylamino)carbonyl]- (9CI, ACI), 2-[(Phenylamino)carbonyl]benzoic acid (ACI), Phthalanilic acid (6CI, 7CI, 8CI), 2-(Anilinocarbonyl)benzoic acid, 2-(Phenylcarbamoyl)benzoic acid, 2-Carboxybenzanilide, Lemax, N-Phenylphthalamic acid, N-Phenylphthalamidic acid, Nevirol, NSC 26414, o-(Phenylcarbamoyl)benzoic acid, Phthalic acid monoanilide, Phthalic monoanilide, Phthalomonoanilide
IUPAC Name 2-(phenylcarbamoyl)benzoic acid
SMILES OC(=O)c1ccccc1C(=O)Nc2ccccc2

Specifications

Quantity 10 g
InChI Formula InChI=1S/C14H11NO3/c16-13(15-10-6-2-1-3-7-10)11-8-4-5-9-12(11)14(17)18/h1-9H,(H,15,16)(H,17,18)
Formula Weight 241.0739
Chemical Name or Material Phthalanilic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.