missing translation for 'onlineSavingsMsg'
Learn More

Phthalan, 98%, Thermo Scientific Chemicals

Product Code. 11419552
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4'-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4′-Di-tert-butyl­biphenyl­, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.

Solubility
Low aqueous solubility.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 496-14-0
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005932
InChI Key SFLGSKRGOWRGBR-UHFFFAOYSA-N
Synonym phthalan, 1,3-dihydroisobenzofuran, isocoumaran, isobenzofuran, 1,3-dihydro, o-xylylene oxide, 2-oxaindan, 1,3-dihydro isobenzofuran, unii-2r6ntq7y6g, ccris 1557, 2r6ntq7y6g
PubChem CID 10327
IUPAC Name 1,3-dihydro-2-benzofuran
SMILES C1C2=CC=CC=C2CO1

Specifications

Density 1.094
Boiling Point 192°C
Flash Point 63°C (145°F)
Refractive Index 1.546
Quantity 5 g
Beilstein 111932
Solubility Information Low aqueous solubility.
Formula Weight 120.15
Percent Purity 98%
Chemical Name or Material Phthalan

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'