Learn More
Phthalan, 98%
Description
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4'-Di-tert-butylbiphenyl, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phthalan is used in the synthesis of nine-membered cyclic dione ethers. It is used along with excess Li metal in the presence of 4,4′-Di-tert-butylbiphenyl, B21470, results in opening of the furan ring. It is used in the synthesis of functionalized benzylic alcohols.
Solubility
Low aqueous solubility.
Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications
Specifications
| Density | 1.094 |
| Boiling Point | 192°C |
| Flash Point | 63°C (145°F) |
| Refractive Index | 1.546 |
| Quantity | 25 g |
| Beilstein | 111932 |
| Solubility Information | Low aqueous solubility. |
| Formula Weight | 120.15 |
| Percent Purity | 98% |
| Chemical Name or Material | Phthalan |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.