missing translation for 'onlineSavingsMsg'
Learn More

Phthalamide, 97%

Product Code. 11372596
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372596 10 g 10g
11382596 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372596 Supplier Thermo Scientific Alfa Aesar Supplier No. L08600.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 88-96-0 | C8H8N2O2 | 164.164 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 88-96-0
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00025478
InChI Key NAYYNDKKHOIIOD-UHFFFAOYSA-N
Synonym phthalamide, 1,2-benzenedicarboxamide, phthaldiamide, phthalic acid diamide, o-carbamoylbenzamide, o-phthalic acid diamide, o-phthalamide, phthalic diamide, ccris 518, unii-7b96053wrs
PubChem CID 6956
ChEBI CHEBI:38799
IUPAC Name benzene-1,2-dicarboxamide
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)N

Tekniske data

Melting Point 223°C (decomposition)
Quantity 10 g
Beilstein 1868220
Merck Index 14,7369
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material Phthalamide

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.