missing translation for 'onlineSavingsMsg'
Learn More

Phthalamic Acid, TRC

Product Code. 30138351
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138351 5 g 5g
30135513 10 g 10g
17981245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138351 Supplier Toronto Research Chemicals Supplier No. TRCP3842505G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-97-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
Synonym Phthalamic acid (6CI,7CI,8CI), 2-(Aminocarbonyl)benzoic acid, 2-Carbamoylbenzoic acid, NSC 5344, Phthalamide acid, Phthalamidic acid, Phthalic acid monoamide, Phthalic monoamide, o-Carbamoylbenzoic acid, Phthalamic Acid, 2-(Aminocarbonyl)benzoic Acid, 2-Carbamoylbenzoic Acid, NSC 5344, Phthalamide Acid, Phthalamidic Acid, Phthalic Acid Monoamide, Phthalic Monoamide, o-Carbamoylbenzoic Acid
IUPAC Name 2-carbamoylbenzoic acid
SMILES NC(=O)c1ccccc1C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H7NO3/c9-7(10)5-3-1-2-4-6(5)8(11)12/h1-4H,(H2,9,10)(H,11,12)
Formula Weight 165.0426
Chemical Name or Material Phthalamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.