missing translation for 'onlineSavingsMsg'
Learn More

Phthalamic acid, 99%

Product Code. 11373156
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373156 25 g 25g
11363156 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373156 Supplier Thermo Scientific Alfa Aesar Supplier No. L12432.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 88-97-1 | C8H7NO3 | 165.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-97-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.148
MDL Number MFCD00025476
InChI Key CYMRPDYINXWJFU-UHFFFAOYSA-N
Synonym phthalamic acid, phthalamidic acid, phthalamide acid, phthalic monoamide, phthalic acid monoamide, o-carbamoylbenzoic acid, benzoic acid, 2-aminocarbonyl, benzoic acid, o-carbamoyl, unii-v344h4pf3y, 2-aminocarbonyl benzoic acid
PubChem CID 6957
ChEBI CHEBI:50736
IUPAC Name 2-carbamoylbenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)N)C(=O)O

Specifications

Melting Point 140°C to 143°C
Quantity 25 g
Beilstein 1868205
Formula Weight 165.15
Percent Purity 99%
Chemical Name or Material Phthalamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.