missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHT-427, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | 4-Dodecyl-N-1,3,4-thiadiazol-2-ylbenzenesulfonamide |
| CAS | 1191951-57-1 |
| Molecular Formula | C20H31N3O2S2 |
| Quantity | 25 mg |
| Synonym | PHT-427, 4-Dodecyl-N-1,3,4-thiadiazol-2-ylbenzenesulfonamide |
| InChI Formula | InChI=1S/C20H31N3O2S2/c1-2-3-4-5-6-7-8-9-10-11-12-18-13-15-19(16-14-18)27(24,25)23-20-22-21-17-26-20/h13-17H,2-12H2,1H3,(H,22,23) |
| SMILES | CCCCCCCCCCCCc1ccc(cc1)S(=O)(=O)Nc2nncs2 |
| IUPAC Name | 4-dodecyl-N-(1,3,4-thiadiazol-2-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 409.61 |
| Formula Weight | 409.1858 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction