missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHT-427, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
PHT-247 is an inhibitor of the pleckstrin homology (PH) domain of Akt, and it is also an inhibitor of PDPK1 with Kis of 2.7 μM and 5.2 μM and for Akt and PDPK1, respectively.
Specifications
Specifications
| Chemical Name or Material | PHT-427 |
| CAS | 1191951-57-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H31N3O2S2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 50 mg/mL (122.07 mM; Need ultrasonic) |
| SMILES | CCCCCCCCCCCCC1=CC=C(C=C1)S(NC2=NN=CS2)(=O)=O |
| Molecular Weight (g/mol) | 409.61 |
| Formula Weight | 409.61 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction