missing translation for 'onlineSavingsMsg'
Learn More

PHT-427, MedChemExpress

Product Code. 16401036
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16401036

Brand: Medchem Express HY12063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PHT-247 is an inhibitor of the pleckstrin homology (PH) domain of Akt, and it is also an inhibitor of PDPK1 with Kis of 2.7 μM and 5.2 μM and for Akt and PDPK1, respectively.

PHT-247 is an inhibitor of the pleckstrin homology (PH) domain of Akt, and it is also an inhibitor of PDPK1 with Kis of 2.7 μM and 5.2 μM and for Akt and PDPK1, respectively.

Specifications

Chemical Name or Material PHT-427
CAS 1191951-57-1
Color White
Purity Grade Notes Research
Molecular Formula C20H31N3O2S2
Quantity 200 mg
Solubility Information DMSO : 50 mg/mL (122.07 mM; Need ultrasonic)
SMILES CCCCCCCCCCCCC1=CC=C(C=C1)S(NC2=NN=CS2)(=O)=O
Molecular Weight (g/mol) 409.61
Formula Weight 409.61
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.