missing translation for 'onlineSavingsMsg'
Learn More

PHRF1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18387292
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387292 100 μg 100µL
18353523 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18387292 Supplier Bio-Techne Supplier No. NBP315531100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PHRF1 Polyclonal antibody specifically detects PHRF1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHRF1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 50% glycerol, pH7.3
Gene Alias CTD binding SR like protein rA9, CTD-binding SR-like protein rA9, KIAA1542RNF221, PHD and RING finger domain-containing protein 1, PHD and ring finger domains 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1350-1450 of human PHRF1 (NP_001273512.1).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 57661
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.