missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHPT1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18413UV
This item is not returnable.
View return policy
Description
PHPT1 Polyclonal antibody specifically detects PHPT1 in Human samples. It is validated for Western Blot
Specifications
| PHPT1 | |
| Polyclonal | |
| Western Blot | |
| bA216L13.10, DKFZp564M173, EC 3.1.3,14 kDa phosphohistidine phosphatase, EC 3.1.3.-, HSPC141, phosphohistidine phosphatase 11700008C22Rik, phosphohistidine phosphatase 14kDa, PHP14CGI-202, Protein janus-A homolog, RP11-216L13.10, sex-regulated protein janus-a | |
| Recognizes region between residues Between between 1 and 50.. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 29085 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction