missing translation for 'onlineSavingsMsg'
Learn More

PHOX2B Rabbit anti-Human, Mouse, Clone: 4W5J8, Novus Biologicals™

Product Code. 18368382
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368382 100 μg 100µL
18378546 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18368382 Supplier Bio-Techne Supplier No. NBP316643100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

PHOX2B Monoclonal antibody specifically detects PHOX2B in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PHOX2B
Applications Western Blot
Classification Monoclonal
Clone 4W5J8
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 215-314 of human PHOX2B (Q99453).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8929
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.