missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2B Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41179H
This item is not returnable.
View return policy
Description
PHOX2B Polyclonal antibody specifically detects PHOX2B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
Specifications
| PHOX2B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA | |
| HRP | |
| NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein | |
| Antibody was raised against a 15 amino acid synthetic peptide near the center of human PHOX2B. The immunogen is located within amino acids 130 - 180 of PHOX2B. Amino Acid Squence: FQRNRRAKFRKQERAA | |
| 0.1 mL | |
| Neuroscience | |
| 8929 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction