missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | PHOX2B |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18648318
|
Novus Biologicals
NBP2-41179-0.025mg |
0.025 mg |
£134.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18109948
|
Novus Biologicals
NBP2-41179 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PHOX2B Polyclonal specifically detects PHOX2B in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| PHOX2B | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat | |
| NBPHOX, NBPhoxPhox2b, Neuroblastoma Phox, paired mesoderm homeobox 2b, paired mesoderm homeobox protein 2B, paired-like homeobox 2bNBLST2, PHOX2B homeodomain protein, PMX2Bneuroblastoma paired-type homeobox protein | |
| PHOX2B | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| PBS with 0.02% Sodium Azide | |
| 8929 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the center of human PHOX2B. The immunogen is located within amino acids 130 - 180 of PHOX2B. Amino Acid Squence: FQRNRRAKFRKQERAA | |
| Primary | |
| PHOX2B antibody is predicted to not cross-react with other paired homeobox family members. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title