missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PHOX2A Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566785
This item is not returnable.
View return policy
Description
Immunogen sequence: AAELLKAWQPA ESGPGPFSGV LSSFHRKPGP ALKTNLF Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.
PHOX2A and PHOX2B (Paired mesoderm homeobox protein) are closely related, paired-homeodomain transcription factors that function as determinants of the noradrenergic phenotype during embryogenesis. PHOX2 proteins are crucial for the regulation of endogenous hydroxylases in neural crest cells and promote sympathetic neuron generation. Human PHOX2B contains one DNA binding homeobox domain and is required for the differentiation of all central and nonperipheral noradrenergic centers in the brain. In contrast, PHOX2A controls only the differentiation of the main noradrenergic center of the brain. Regulation of PHOX2 may have therapeutic utility in aging or disorders involving degeneration of noradrenergic neurons.
Specifications
| PHOX2A | |
| Polyclonal | |
| Unconjugated | |
| PHOX2A | |
| aristaless homeobox gene homolog; aristaless homeobox homolog; aristaless homeobox protein homolog; ARIX; arix homeodomain protein; ARIX1 homeodomain protein; CFEOM2; FEOM2; NCAM2; paired like homeobox 2a; paired mesoderm homeobox 2a; paired mesoderm homeobox protein 2A; Paired-like homeobox 2A; Phox2; PHOX2A; PHOX2A homeodomain protein; Pmx2; Pmx2a | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 401 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14813 | |
| PHOX2A | |
| Recombinant Human PHOX2A. Recombinant protein control fragment (Product #RP-107444). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction