missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHOX2A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41169
This item is not returnable.
View return policy
Description
PHOX2A Polyclonal specifically detects PHOX2A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| PHOX2A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| PHOX2A | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human PHOX2A. The immunogen is located within the last 50 amino acids of PHOX2A. Amino Acid Squence: ELLKAWQPAESGPGPF | |
| 0.1 mg | |
| Growth and Development, Neuronal Cell Markers, Neuroscience | |
| 401 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| aristaless (Drosophila) homeobox, aristaless homeobox (Drosophila), fibrosis ofextraocular muscles, congenital, 2, autosomal recessive, aristaless homeobox homolog, Aristaless homeobox protein homolog, arix homeodomain protein, ARIX1 homeodomain protein, ARIXNCAM2, CFEOM2MGC52227, paired mesoderm homeobox protein 2A, paired-like homeobox 2apaired-like (aristaless) homeobox 2a, PMX2AFEOM2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| PHOX2A antibody is predicted to not cross-react with other paired homeobox family members. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction