missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Photometry Test Reagent
missing translation for 'orderingAttributeHoverText'
Quantity:
100 Pack
15 mL
Type:
Triazole Reagent, F25
Urea Reagent 2
missing translation for 'unitSize'
15mL
Pack of 100
Description
- Highest quality production standards
- Maximum accuracy
Specifications
Specifications
| Format | Liquid |
| Quantity | 15 mL |
| Type | Urea Reagent 2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction