Learn More
Palintest™ Photometer Tablet Reagents
Product Code. 11913657
Click to view available options
Range:
0 to 0.5mg/L
0 to 1.5mg/L
0 to 100mg/L
0 to 2.5mg/L
0 to 20mg/L
0 to 250mg/L
0 to 500mg/L
2.5 to 2mg/L
Type:
Alkalinity Total (Alkaphot) Test Kit
Chlorine Dioxide HR Test Kit
Chlorine Dioxide LR Test Kit
Chlorine HR Test Kit
Fluoride Test Kit
Molybdate HR Test Kit
Organophosphonate (OP) Test Kit
Sulphide Test Kit
Unit Size:
50 tests
Each
Pack of 50
Product Code. 11913657
Brand: Palintest™ PM168
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
  • Convenient tablet form.
  • Easy to use.
  • Available for a variety of water testing needs.
Specifications
No. of Tests 50
Quantity 50/Pk.
Range 0 to 0.5mg/L
Sufficient For 50 Tests
Type Sulphide Test Kit