missing translation for 'onlineSavingsMsg'
Learn More

Photo-lysine, MedChemExpress

Product Code. p-200101871
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
20 mg
Unit Size:
10mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 16470378

Brand: Medchem Express HY19804

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Photo-lysine, a new lysine-based photo-reactive amino acid, captures proteins that bind lysine post-translational modifications.

Photo-lysine, a new lysine-based photo-reactive amino acid, captures proteins that bind lysine post-translational modifications.

Specifications

Chemical Name or Material Photo-lysine
CAS 1863117-91-2
Purity Grade Notes Research
Molecular Formula C6H12N4O2
Synonym Photo lysine
SMILES NCCC1(N=N1)C[C@@H](C(O)=O)N
Molecular Weight (g/mol) 172.19
Quantity 5 mg
Formula Weight 172.19
Grade Research

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.