missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Phosphotyrosine Monoclonal Antibody (4B6C8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA538227
This item is not returnable.
View return policy
Description
Positive Samples: Jurkat.
The role of tyrosine phosphorylation in transduction of the mitogenic signal from transmembrane receptors and in transformation by oncogene tyrosine kinases has been the subject of intense investigation for several years. While the phosphorylation of specific tyrosine residues has been shown to be a primary mechanism of signal transduction during normal mitogenesis, cell cycle progression and oncogenic transformation, its role in other areas such as differentiation and gap junction communication, is a matter of active and ongoing research. Antibodies that specifically recognize phosphorylated tyrosine residues have proved to be invaluable to the study of tyrosine -phosphorylated proteins and the biochemical pathways in which they function. The fluorescein (FITC) conjugate of clone PY20 anti-phosphotyrosine is especially useful for the detection of these P-Tyr proteins in immunohistochemical and immunocytochemical protocols in situations wherein the use of a secondary antibody would complicate detection of the protein(s) of interest.
Specifications
| Phosphotyrosine | |
| Monoclonal | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| Chemical, Human, Mouse, Rat | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot, Immunocytochemistry | |
| 4B6C8 | |
| Unconjugated | |
| Phosphotyrosine; pTyr; pY | |
| A synthetic peptide corresponding to a sequence containing phosphorylated Y. | |
| 100 μL | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction