missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic Acid Hydrate, MP Biomedicals™

Product Code. 11483612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483612 500 g 500g
11453612 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483612 Supplier MP Biomedicals Supplier No. 0215624390
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphotungstic Acid Hydrate is used for cell staining.

  • Phosphotungstic acid is used in histology as a component for staining of cell specimens.
  • It is a common negative stain for viruses, nerves, polysaccharides, and other biological tissue materials for imaging by a transmission electron microscope.
  • Phosphotungstic acid has been used to precipitate different types of dyes as lakes.

Identificatori chimici

CAS 12501-23-4
Molecular Formula H5O41PW12
Molecular Weight (g/mol) 2880.20
MDL Number MFCD00011342
InChI Key AVFBYUADVDVJQL-UHFFFAOYSA-N
Synonym phosphotungstic acid hydrate, tungstophosphoric acid, 12-tungstophosphoric acid hydrate, phosphoric acid dodecakis tungsten trioxide hydrate, iuaxea, aronis24073, phosphotungstic acid hydrate, reagent grade, phosphotungstic acid hydrate, for microscopy, phosphotungstic acid hydrate, p.a, phosphotungstic acid hydrate, saj special grade
PubChem CID 16212977
IUPAC Name 12-Tungstophosphate hydrate
SMILES O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.OP(O)(O)=O

Specifica

Color White
Melting Point ∼95°C
Quantity 500 g
Physical Form Crystalline Powder
Solubility Information Soluble in water (1 g/10 mL), alcohol and ether.
Formula Weight 2880.2 (anhyd)

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.