missing translation for 'onlineSavingsMsg'
Learn More

Phosphotungstic acid hydrate, Thermo Scientific Chemicals

Product Code. 11359247
Click to view available options
Quantity:
25 g
100 g
500 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phosphotungstic acid is used as a catalyst for the hydrolysis of propene to prepare 2-propanol. It is also involved in heterogeneous catalysis reaction to prepare propene from 2-propanol and methanol from hydrocarbons. Its ethanolic
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12067-99-1
MDL Number MFCD00041964
Synonym Tungstophosphoric acid

Specifications

Color White
Odor Odorless
Quantity 100 g
Molecular Formula P2O5·24WO3·xH2O
Physical Form Powder
UN Number UN3260
Merck Index 14,7364
Solubility Information Soluble in water, alcohol and ether
Formula Weight 5706.35 (Anhydrous)
Chemical Name or Material Phosphotungstic acid hydrate
compliance-icons
Product Identifier
  • Phosphotungstic acid hydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'