missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoserine phosphatase Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90222-50UL
This item is not returnable.
View return policy
Description
Phosphoserine phosphatase Monoclonal specifically detects Phosphoserine phosphatase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| Phosphoserine phosphatase | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence, Immunoprecipitation | |
| 1 | |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 0.2-1 ul/mg of lysate, Immunofluorescence | |
| L-3-phosphoserine phosphatase, O-phosphoserine phosphohydrolase, phosphoserine phosphatase, PSPase, PSPEC 3.1.3.3 | |
| Recombinant Human Phosphoserine phosphatase protein | |
| 50 μg | |
| Lipid and Metabolism | |
| 5723 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction