missing translation for 'onlineSavingsMsg'
Learn More

Phosphoserine phosphatase Antibody, Novus Biologicals™

Product Code. 30232281 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30232281 100 μL 100µL
30231980 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30232281 Supplier Novus Biologicals Supplier No. NBP333385100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Phosphoserine phosphatase Monoclonal antibody specifically detects Phosphoserine phosphatase in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoserine phosphatase
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, ELISA Recommended starting concentration is 1 μg/mL
Formulation PBS (pH 7.3), 50% glycerol, 0.05% BSA
Gene Alias L-3-phosphoserine phosphatase, O-phosphoserine phosphohydrolase, phosphoserine phosphatase, PSPase, PSPEC 3.1.3.3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-225 of human Phosphoserine phosphatase (NP_004568.2).,, Sequence:, MVSHSELRKLFYSADAVCFDVDSTVIREEGIDELAKICGVEDAVSEMTRRAMGGAVPFKAALTERLALIQPSREQVQRLIAEQPPHLTPGIRELVSRLQERNVQVFLISGGFRSIVEHVASKLNIPATNVFANRLKFYFNGEYAGFDETQPTAESGGKGKVIKLLKEKFHFKKIIMIGDGATDMEACPPADAFIGFGGNVIRQQVKDNAKWYITDFVELLGELEE
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5723
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.