missing translation for 'onlineSavingsMsg'
Learn More

Phosphoserine phosphatase Antibody (3G12), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18428887 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428887 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428887 Supplier Novus Biologicals Supplier No. NBP104297JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phosphoserine phosphatase Monoclonal antibody specifically detects Phosphoserine phosphatase in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphoserine phosphatase
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 3G12
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias L-3-phosphoserine phosphatase, O-phosphoserine phosphohydrolase, phosphoserine phosphatase, PSPase, PSPEC 3.1.3.3
Host Species Mouse
Immunogen Recombinant human Phosphoserine phosphatase (1-225aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5723
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.