missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoserine phosphatase Antibody (3G12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-04297
This item is not returnable.
View return policy
Description
Phosphoserine phosphatase Monoclonal specifically detects Phosphoserine phosphatase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Phosphoserine phosphatase | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:2000, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
| L-3-phosphoserine phosphatase, O-phosphoserine phosphohydrolase, phosphoserine phosphatase, PSPase, PSPEC 3.1.3.3 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Anti-human PSP mAb, clone 3G12, is derived from hybridization of mouse SP2/O myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PSP protein. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 3G12 | |
| Unconjugated | |
| NP_004568 | |
| PSPH | |
| Recombinant human PSP (1-225aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5723 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction