missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phosphoserine Monoclonal Antibody (106.1)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA191800

Product Code. 11565662

  • £479.00 / 500µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: Brain Cortex.

Phosphorylated serine (phosphoserine) is an alpha amino acid that together with phosphothreonine, is one of the most ubiquitously modified amino acids in proteins. Many hormones, such as adrenaline (epinephrine), alter the activities of enzymes by stimulating the phosphorylation of serine and threonine, and these can have profound effects on cellular signal transduction, apoptosis and carcinogenesis.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phosphoserine
Monoclonal
0.65 mg/mL
PBS with 0.2% BSA and 0.1% sodium azide
Mouse
Protein A
RUO
Chemical
Antibody
IgG1
Western Blot
106.1
Unconjugated
C3H8NO6P; Dexfosfoserine; Fosforina; L-O-Phosphoserine; O-phospho-L-serine; O-Phosphoserine; Phospho-Serine; pSer
Phosphoserine conjugated to KLH.
500 μL
Primary
4°C, do not freeze
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.