missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoserine Antibody (PSR-45), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£175.00
Specifications
| Antigen | Phosphoserine |
|---|---|
| Clone | PSR-45 |
| Dilution | Western Blot 1:500-1:1000, ELISA 1:4000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Phosphoserine Monoclonal specifically detects Phosphoserine in Human, Rat, Rabbit samples. It is validated for Western Blot, ELISA.Specifications
| Phosphoserine | |
| Western Blot 1:500-1:1000, ELISA 1:4000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Phosphoserine; Phosphothreonine | |
| IgG1 | |
| Unpurified | |
| By ELISA and dot blot, the antibody reacts specifically with phosphorylated serine, both as free amino acid or conjugated to carriers such as BSA or KLH. No cross-reactivity is observed with non-phosphorylated serine, phosphothreonine, phosphotyrosine, AMP or ATP. This antibody has been used in immunoblotting for the localization of some phosphoserine-containing proteins. Certain proteins known to contain phosphorylated serine may not be recognized by this antibody due to steric hindrance of the recognition site. |
| PSR-45 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Phosphoserine conjugated to Keyhole Limpet Hemocyanin (KLH). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title