missing translation for 'onlineSavingsMsg'
Learn More

Phosphorylethanolamine, TRC

Product Code. 30012906
Change view
Click to view available options
Quantity:
10 mg
1 g
5 g
Unit Size:
10mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012906 5 g 5g
30007497 10 mg 10mg
17984905 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012906 Supplier Toronto Research Chemicals Supplier No. TRCP3609005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071-23-4
Molecular Formula C2 H8 N O4 P
Molecular Weight (g/mol) 141.06
Synonym 2-Aminoethanol 1-(dihydrogen phosphate), Ethanol, 2-amino-, dihydrogen phosphate (ester) (8CI,9CI), Ethanol, 2-amino-, phosphate (6CI), Phosphoric acid, 2-aminoethyl ester (6CI), 2-Aminoethanol O-phosphate, 2-Aminoethyl dihydrogen phosphate, Colamine phosphate, Ethanolamine O-phosphate, Mono(2-aminoethyl) phosphate, Monoaminoethyl phosphate, NSC 254167, O-Phosphoethanolamine, O-Phosphorylethanolamine, Phosphoethanolamine, Phosphonoethanolamine, Phosphorylethanolamine
IUPAC Name 2-aminoethyl dihydrogen phosphate
SMILES NCCOP(=O)(O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H8NO4P/c3-1-2-7-8(4,5)6/h1-3H2,(H2,4,5,6)
Formula Weight 141.0191
Percent Purity >95%
Chemical Name or Material Phosphorylethanolamine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.