missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Phosphorylase B Native Protein

Product Code. 18658176 Shop All Bio Techne Products
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 18658176

Brand: Novus Biologicals™ NBP262520

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

Concentration 5 mg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 5257
Molecular Weight (g/mol) MolecularWeight-theroretical: 185 kDa
Quantity 25 mg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias DKFZp781E15103, FLJ41698, phosphorylase b kinase regulatory subunit beta, phosphorylase kinase beta-subunit, Phosphorylase kinase subunit beta, phosphorylase kinase, beta
Gene Symbol PHKB
Reconstitution Reconstitute in sterile distilled water.
Research Category Cancer, Endocrinology, Signal Transduction
Purity or Quality Grade >95%
Protein Phosphorylase B
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.