missing translation for 'onlineSavingsMsg'
Learn More

Phosphorylase B Antibody (2E9), Novus Biologicals™

Product Code. 18356739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356739 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356739 Supplier Novus Biologicals Supplier No. H00005257M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Phosphorylase B Monoclonal antibody specifically detects Phosphorylase B in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phosphorylase B
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2E9
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_000284
Gene Alias DKFZp781E15103, FLJ41698, phosphorylase b kinase regulatory subunit beta, phosphorylase kinase beta-subunit, Phosphorylase kinase subunit beta, phosphorylase kinase, beta
Host Species Mouse
Immunogen PHKB (NP_000284, 984 a.a. ~ 1093 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. EDTLGNIDQPQYRQIVVELLMVVSIVLERNPELEFQDKVDLDRLVKEAFNEFQKDQSRLKEIEKQDDMTSFYNTPPLGKRGTCSYLTKAVMNLLLEGEVKPNNDDPCLIS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5257
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.