missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxybromide, 98% min

Product Code. p-3208576
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11488660

Brand: Thermo Scientific Alfa Aesar 056135.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Use As a Chemical Intermediate | CAS: 7789-59-5 | Br3OP | 286.685 g/mol

As a chemical intermediate.Phosphorus(V) oxybromide is used as a brominating agent in the synthesis of 2,4,6-tribromopyrimidine and bromoheterocyclic compounds. It is used in the phosphorus flame retardant and phosphorus activators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-59-5
Molecular Formula Br3OP
Molecular Weight (g/mol) 286.685
MDL Number MFCD00036298
InChI Key UXCDUFKZSUBXGM-UHFFFAOYSA-N
Synonym phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
PubChem CID 24613
SMILES O=P(Br)(Br)Br

Specifications

Boiling Point 189°C
Melting Point 56°C
Color White to Yellow
Physical Form Lump
Quantity 5 g
Assay Percent Range 98% min.
Solubility Information Decomposes in water. Soluble in ether,carbon disulfide,benzene,chloroform and sulfuric acid.
Formula Weight 286.73
Sensitivity Moisture sensitive
Density 2.822
Chemical Name or Material Phosphorus(V) tribromide oxide
Show More Show Less
compliance-icons
Product Identifier
  • Phosphorus(V) oxybromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Phosphorus oxybromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.