missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxide, Puratronic™, 99.9925% (metals basis), Thermo Scientific Chemicals

Product Code. p-7048544
Click to view available options
Quantity:
100 g
5 x 100 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11306967

Brand: Thermo Scientific Alfa Aesar 010827.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A dehydrating agent often used as a desiccant for bacterial cultural extracts analyzed by gas-liquid chromatography. Used for the preparation of phosphoric acid, phosphorus compound, it can also be used for the preparation of high-purity phosphoric acid. Also used in sugar refining and a catalyst for air blowing of asphalt, reagent in the efficient and modified Biginelli-type synthesis of 3,4-dihydro-1H-indeno[1,2-d]pyrimidine-2,5-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Melting Point 340°C
Density 2.39 g/mL
Boiling Point 360°C (sublimation)
Odor Weak
Quantity 100 g
UN Number UN1807
Sensitivity Moisture sensitive
Merck Index 14,7355
Solubility Information Soluble in sulfuric acid. Insoluble in acetone and ammonia.
Formula Weight 141.94
Percent Purity 99.9925%
Grade Puratronic
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Phosphorus(V) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.