missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxide, 99.99%, Thermo Scientific Chemicals

Catalog Number 11433880
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
1314-56-3
O5P2
141.94
MFCD00011440
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
This item is not returnable. View return policy
1314-56-3
O5P2
141.94
MFCD00011440
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Phosphorus(V) oxide is used as a drying and dehydrating agent, a condensation reagent in organic synthesis and a laboratory reagent. It is also used in sugar refining and in fire extinguishing. Phosphorus pentoxide in DMSO forms an Onodera reagent which oxidizes alcohols. It is capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 360°C (sublimation)
Melting Point 340°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99%
Solubility Information Soluble in sulfuric acid. Insoluble in acetone and ammonia. Decomposes in water.
Formula Weight 141.94
Odor Odorless
Sensitivity Moisture sensitive
Density 2.39 g/cm3 at 20°C
Chemical Name or Material Phosphorus(V) oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'