missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxide, 99.99%

Product Code. 11423880
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423880 100 g 100g
11433880 10 g 10g
11413880 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423880 Supplier Thermo Scientific Alfa Aesar Supplier No. 089966.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1314-56-3 | O5P2 | 141.94 g/mol

Phosphorus(V) oxide is used as a drying and dehydrating agent, a condensation reagent in organic synthesis and a laboratory reagent. It is also used in sugar refining and in fire extinguishing. Phosphorus pentoxide in DMSO forms an Onodera reagent which oxidizes alcohols. It is capable of converting mineral acids to anhydrides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-56-3
Molecular Formula O5P2
Molecular Weight (g/mol) 141.94
MDL Number MFCD00011440
InChI Key DLYUQMMRRRQYAE-UHFFFAOYSA-N
IUPAC Name tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
SMILES O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

Specifications

Boiling Point 360°C (sublimation)
Melting Point 340°C
Color White
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.99%
Solubility Information Soluble in sulfuric acid. Insoluble in acetone and ammonia. Decomposes in water.
Formula Weight 141.94
Odor Odorless
Sensitivity Moisture sensitive
Density 2.39 g/cm3 at 20°C
Chemical Name or Material Phosphorus(V) oxide
Show More Show Less
compliance-icons
Product Identifier
  • Phosphorus(V) oxide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phosphorus oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.