missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus trichloride, 99%, Thermo Scientific Chemicals

Catalog Number 10397390
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-12-2
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-12-2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 5 g
Assay Percent Range 99%
Molecular Formula Cl5Ta
Linear Formula PCl3
MDL Number MFCD00011253
Fieser 01,875; 08,400; 10,318
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]
IUPAC Name tantalum(5+) pentachloride
Molecular Weight (g/mol) 358.20
Formula Weight 137.33
Percent Purity ≥98.5%
Packaging Glass bottle
Density 1.5700g/mL
Chemical Name or Material Phosphorus trichloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus trichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H300-Fatal if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'