missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Standard: P @ 1 wt% (10,000μg/g) in 75 cSt Hydrocarbon Oil

Product Code. 30322352
Change view
Click to view available options
Quantity:
400 g
Unit Size:
400g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30322352 400 g 400g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30322352 Supplier VHG Labs Supplier No. VHGOP1P400G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorus
CAS 7723-14-0
Analyte or Component Names Phosphorus
Shipping Condition Room Temperature
Molecular Formula P
Concentration 1 %
Synonym P
InChI Formula InChI=1 S/P
Solution Type Single Solution
SMILES [P]
Element P
IUPAC Name phosphorus
Solvent or Matrix 75 cSt Hydrocarbon Oil
Industry Type Industrial QC
Molecular Weight (g/mol) 30.97
Quantity 400 g
Formula Weight 30.9738
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.