missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus Solution for ICP, 1g/L, H2O Matrix, Fisher Chemical™
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
£52.40 - £114.00
Specifications
Concentration | 1g/L |
---|---|
Solution Contains | H2O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12973464
|
Fisher Chemical
S 040 100 |
100 mL |
£52.40
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12983464
|
Fisher Chemical
S 040 250 |
250 mL |
£79.90
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12993464
|
Fisher Chemical
S 040 500 |
500 mL |
£114.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
1g/L | |
H2O |