missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus Pentasulfide, 99%, Honeywell™ Fluka™

Product Code. 15612310 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
1 kg
Packaging:
Glass Bottle
Unit Size:
100g
1kg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15612310 5 g Glass Bottle 5g
15692300 100 g Glass Bottle 100g
15602310 1 kg Glass Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15612310 Supplier Honeywell-Fluka Supplier No. 2321065G
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 1314-80-3
Molecular Formula P4S10
Molecular Weight (g/mol) 444.495
MDL Number MFCD00011441
InChI Key CYQAYERJWZKYML-UHFFFAOYSA-N
Synonym phosphorus pentasulfide, sulfur phosphide, diphosphorus pentasulfide, diphosphorus pentasulphide, tetraphosphorus decasulfide, phosphoric sulfide, phosphorus persulfide, phosphorus pentasulphide, thiophosphoric anhydride, sirnik fosforecny czech
PubChem CID 14817
SMILES P12(=S)SP3(=S)SP(=S)(S1)SP(=S)(S2)S3

Specifikationer

Boiling Point 515°C
Melting Point 280°C to 284°C
Color Green-Yellow to Yellow
Quantity 5 g
UN Number UN1340
Formula Weight 222.27g/mol
Percent Purity 99%
Packaging Glass Bottle
Appearance Powder
Density 2.09g/mL at 25°C
Chemical Name or Material Phosphorus Pentasulfide
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.