missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus oxychloride, 99%, Thermo Scientific Chemicals

Product Code. 10133780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
Product Code. Quantity Packaging unitSize
10133780 1 kg Glass bottle 1kg
1 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10133780

Brand: Thermo Scientific Acros 191290010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-87-3
MDL Number MFCD00011443
InChI Key XHXFXVLFKHQFAL-UHFFFAOYSA-N
Synonym phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
PubChem CID 24813
ChEBI CHEBI:30336
SMILES O=P(Cl)(Cl)Cl

Specifications

Boiling Point 107.0°C
Melting Point 1.2°C
Physical Form Liquid
Quantity 1 kg
Assay Percent Range 98.5% min. (ex Cl) (Argentometry)
Molecular Formula Cl3OP
Linear Formula POCl3
Fieser 01,876; 02,330; 03,228; 04,390; 05,535; 07,292; 09,374; 11,429; 13,249; 15,267; 17,288
Merck Index 15, 7461
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ccl4, chloroform and benzene
Molecular Weight (g/mol) 153.33
Formula Weight 153.33
Percent Purity 99%
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620
Specific Gravity 1.645
Density 1.6450g/mL
Chemical Name or Material Phosphorus oxychloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus oxychloride
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H372-Causes damage to organs.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.