missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(III) fluoride, 99%

Product Code. 15481537
Click to view available options
Quantity:
50 g
Packaging:
Ampule
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 15481537

Brand: Thermo Scientific Alfa Aesar 039525.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7783-55-3 | F3P | 87.969 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point -101.5°C
Melting Point -151.5°C
CAS 7783-55-3
Color Colorless
Physical Form Gas
Quantity 50 g
Molecular Formula F3P
MDL Number MFCD00040325
UN Number UN1955
Merck Index 14,7359
Synonym phosphorus trifluoride, trifluorophosphine, phosphorous fluoride, phosphorous trifluoride, phosphorus fluoride, trifluorophosphin, unii-496073dybf, phosphorus iii fluoride, pf3, trifluoridophosphorus
InChI Key WKFBZNUBXWCCHG-UHFFFAOYSA-N
SMILES FP(F)F
IUPAC Name trifluorophosphane
Molecular Weight (g/mol) 87.969
PubChem CID 62665
ChEBI CHEBI:30205
Formula Weight 87.97
Percent Purity 99%
Odor Odorless
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.907 g/mL
Chemical Name or Material Phosphorus(III) fluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.