missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus, AAS standard solution, Specpure™ P 1000μg/mL

Product Code. 11306409
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11306409 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11306409 Supplier Thermo Scientific Alfa Aesar Supplier No. 089887.AP
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7723-14-0 | P | 30.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7723-14-0, 7697-37-2, 7732-18-5
Molecular Formula P
MDL Number MFCD00133771
InChI Key OAICVXFJPJFONN-UHFFFAOYSA-N
SMILES [P]
IUPAC Name phosphorus
Molecular Weight (g/mol) 30.97
PubChem CID 5462309
Quantity 500 mL
ChEBI CHEBI:30207
Chemical Name or Material Phosphorus
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.