missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(III) bromide, 99%

Product Code. 11399327
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399327 25 g 25g
11389327 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399327 Supplier Thermo Scientific Alfa Aesar Supplier No. 040191.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-60-8 | Br3P | 270.686 g/mol

Phosphorus(III) bromide is used mainly for the conversion of alcohols to alkyl bromides and it is used in tests for skin corrosivity. It is used for alkene preparation and it act as a lewis base and a lewis acid. It is used as a catalyst for the alpha-bromination of carboxylic acids and used as intermediates in hell-volhard-zelinsky halogenation. Phosphorus tribromide is used in the manufacture of pharmaceuticals such as alprazolam, methohexital and fenoprofen. It is also used as a fire suppression agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-60-8
Molecular Formula Br3P
Molecular Weight (g/mol) 270.686
MDL Number MFCD00011436
InChI Key IPNPIHIZVLFAFP-UHFFFAOYSA-N
Synonym phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
PubChem CID 24614
IUPAC Name tribromophosphane
SMILES P(Br)(Br)Br

Specifications

Boiling Point 172.9°C
Melting Point °C to 40°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99% (metals basis)
Beilstein 3903060
Solubility Information Micible in acetone,carbon disulfide,hydrogen sulfide,chloroform,diethyl ether and decomposes in water and alcohol.
Formula Weight 270.7
Odor Pungent
Refractive Index 1.697
Sensitivity Moisture sensitive
Appearance Colorless liquid
Density 2.852 g/cm3 at 20°C
Chemical Name or Material Phosphorus(III) bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Phosphorus(III) bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Phosphorus tribromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.