missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(III) bromide, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-3208574
Click to view available options
Quantity:
25 g
100 g
7789-60-8
Br3P
270.686
MFCD00011436
IPNPIHIZVLFAFP-UHFFFAOYSA-N
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
24614
tribromophosphane
P(Br)(Br)Br
7789-60-8
Br3P
270.686
MFCD00011436
IPNPIHIZVLFAFP-UHFFFAOYSA-N
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine
24614
tribromophosphane
P(Br)(Br)Br

Phosphorus(III) bromide is used mainly for the conversion of alcohols to alkyl bromides and it is used in tests for skin corrosivity. It is used for alkene preparation and it act as a lewis base and a lewis acid. It is used as a catalyst for the alpha-bromination of carboxylic acids and used as intermediates in hell-volhard-zelinsky halogenation. Phosphorus tribromide is used in the manufacture of pharmaceuticals such as alprazolam, methohexital and fenoprofen. It is also used as a fire suppression agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 172.9°C
Melting Point °C to 40°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Assay Percent Range 99% (metals basis)
Beilstein 3903060
Solubility Information Micible in acetone,carbon disulfide,hydrogen sulfide,chloroform,diethyl ether and decomposes in water and alcohol.
Formula Weight 270.7
Odor Pungent
Refractive Index 1.697
Sensitivity Moisture sensitive
Appearance Colorless liquid
Density 2.852 g/cm3 at 20°C
Chemical Name or Material Phosphorus(III) bromide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'