missing translation for 'onlineSavingsMsg'
Learn More

Phosphorothioic Acid Trisodium Salt Hydrate, TRC

Product Code. 30414295
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30414295 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30414295 Supplier Toronto Research Chemicals Supplier No. TRCP35951525G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphorothioic Acid Trisodium Salt Hydrate
CAS 10489-48-2
Quantity 25 g
Molecular Formula Na3O3PS • x( H2O)
Synonym Sodium Phosphorothioate, Na3PO3S Hydrate
InChI Formula InChI=1S/3Na.H3O3PS.H2O/c;;;1-4(2,3)5;/h;;;(H3,1,2,3,5);1H2/q3*+1;;/p-3
SMILES O=P([O-])([S-])[O-].[Na+].O.[Na+].[Na+]
IUPAC Name sodium phosphorothioate hydrate
Molecular Weight (g/mol) 198.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.