missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, ULTREX™ II Ultrapure Reagent, J.T. Baker™

Product Code. 15528384 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Poly Bottle
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15528384 50 g Poly Bottle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528384 Supplier Avantor J.T.Baker Supplier No. 690804

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagents for critical elemental analysis with less than 10 parts-per-trillion (ppt) levels of up to 65 elements

    For use in ICP (Inducutively Coupled Plasma Spectrometry), Trace Metal Analysis; For drug manufacturing or R&D use only; Not on TSCA Inventory

Specifications

CAS 7664-38-2
Packaging Poly Bottle
Specific Gravity 1L = 1.69kg
Quantity 50 g
Molecular Formula H3O4P
Synonym orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
SMILES OP(=O)(O)O
IUPAC Name phosphoric acid
Molecular Weight (g/mol) 97.994
PubChem CID 1004
ChEBI CHEBI:26078
Formula Weight 98
Grade ULTREX™ II Ultrapure Reagent
Chemical Name or Material Phosphoric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.