missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid Tris(3-methylphenyl) Ester, TRC

Product Code. 30136339
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136339 1 g 1g
30138207 5 g 5g
17981055 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136339 Supplier Toronto Research Chemicals Supplier No. TRCP3594551G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-04-2
Molecular Formula C21 H21 O4 P
Molecular Weight (g/mol) 368.36
Synonym Phosphoric acid, tris(3-methylphenyl) ester (9CI, ACI), m-Tolyl phosphate, (C7H7O)3PO (6CI), Phosphoric acid, tri-m-tolyl ester (8CI), NSC 4055, TMCP, Tri-m-cresyl phosphate, Tri-m-tolyl phosphate, Tris(3-methylphenyl) phosphate, Tris(m-tolyl) phosphate, Tris-m-cresyl phosphate, Tri-3-cresyl phosphate
IUPAC Name tris(3-methylphenyl) phosphate
SMILES Cc1cccc(OP(=O)(Oc2cccc(C)c2)Oc3cccc(C)c3)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C21H21O4P/c1-16-7-4-10-19(13-16)23-26(22,24-20-11-5-8-17(2)14-20)25-21-12-6-9-18(3)15-21/h4-15H,1-3H3
Formula Weight 368.1177
Chemical Name or Material Tri-m-cresyl phosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.